2-Amino-5-bromo-3-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A211125-10G A211125-250MG A211125-1G A211125-25G A211125-5G
103818-082EA 9 USD
103818-082 103848-774 76825-908 76825-910 77087-178 77087-180 77087-182
2-Amino-5-bromo-3-methoxybenzoic acid 98%
2-Amino-5-bromo-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MW: 246.058 g/mol
Boiling Pt: 356 °C (760 mm Hg)
Flash Pt: 169.1 °C
Storage Temperature: Ambient
MDL Number: MFCD11101005
CAS Number: 864293-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR