2-Amino-5-bromo-3-fluorobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromo-2-carboxy-6-fluoroaniline
A131821-250MG
A131821-1G
A131821-25G
103801-640EA
26
USD
103801-640
77095-010
77314-172
77611-168
2-Amino-5-bromo-3-fluorobenzoic acid 97%
2-Amino-5-bromo-3-fluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₅BrFNO₂ MW: 234.023 g/mol Boiling Pt: 321.7±42.0 °C (760 mm Hg) Melting Pt: 217…221 °C Flash Pt: 148.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD04035659 CAS Number: 874784-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...