2-Amino-5-acetylpyridine 95%

Supplier: Ambeed
Warning

A129187-100MG A129187-250MG A129187-1G A129187-5G
103801-240EA 78 USD
103801-240 76971-974 76971-976 77313-604 77313-606
2-Amino-5-acetylpyridine 95%
2-Amino-5-acetylpyridine

Precaución: for Research Use Only

Formula: C₇H₈N₂O
MW: 136.15 g/mol
Boiling Pt: 332.356 °C (760 mm Hg)
Flash Pt: 154.804 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09038484
CAS Number: 19828-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR