2-Amino-5-(trifluoromethyl)benzonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-3-cyanobenzotrifluoride, 2-Amino-5-trifluoromethylbenzonitrile

A241687-1G A241687-25G A241687-100G A241687-10G A241687-250MG
103811-420EA 7 USD
103811-420 103851-224 77090-388 77090-390 77113-254 77113-256 77113-258
2-Amino-5-(trifluoromethyl)benzonitrile 97%
2-Amino-5-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₅F₃N₂
MW: 186.1300 g/mol
Boiling Pt: 268.507 °C (760 mm Hg)
Flash Pt: 116.189 °C
Storage Temperature: Ambient
MDL Number: MFCD03407465
CAS Number: 6526-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR