2-Amino-4-oxo-4,7-dihydro-3H-pyrrolo[2,3-d]pyrimidine-5-carbonitrile 95%
Supplier: Ambeed
|
Warning
|
A152204-100MG
A152204-250MG
103804-362EA
149
USD
103804-362
103842-524
77028-186
77667-154
2-Amino-4-oxo-4,7-dihydro-3H-pyrrolo[2,3-d]pyrimidine-5-carbonitrile 95%
2-Amino-4-oxo-4,7-dihydro-3H-pyrrolo[2,3-d]pyrimidine-5-carbonitrile
Precaución: for Research Use Only
Formula:
C₇H₅N₅O Storage Temperature: Refrigerator |
MDL Number:
MFCD13178502 CAS Number: 69205-79-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...