2-Amino-4-nitrobenzonitrile 97%

Supplier: Ambeed
Warning

A102948-250MG A102948-25G
103796-452EA 17 USD
103796-452 103822-158 77148-322 77148-324 77891-758
2-Amino-4-nitrobenzonitrile 97%
2-Amino-4-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
Boiling Pt: 414.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04117876
CAS Number: 87376-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR