4-Nitroanthranilic acid 98%

Supplier: Ambeed
Warning

A141771-100G A141771-10G A141771-500G A141771-1000G A141771-25G
103808-618EA 231 USD
103808-618 76767-798 76767-800 76767-802 77316-242 77316-244
4-Nitroanthranilic acid 98%
4-Nitroanthranilic acid

Precaución: for Research Use Only

Formula: C₇H₆N₂O₄
Boiling Pt: 431.4 °C (760 mm Hg)
Melting Pt: 257 °C
Flash Pt: 214.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00007262
CAS Number: 619-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR