2-Amino-4-methylnicotinonitrile 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-4-methylpyridine-3-carbonitrile, 2-Amino-3-cyano-4-methylpyridine
A642738-1G
A642738-250MG
103840-352EA
224
USD
103840-352
76844-530
77254-620
2-Amino-4-methylnicotinonitrile 97%
2-Amino-4-methylnicotinonitrile
Precaución: for Research Use Only
Formula:
C₇H₇N₃ MW: 133.151 g/mol Boiling Pt: 306.3 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD09030794 CAS Number: 71493-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...