2-Amino-4-methylbenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-p-toluic acid, 4-Methylanthranilic acid, 4-Methylanthranil acid, 2-Amino-4-methylbenzoic Acid

A203938-10G A203938-25G A203938-5G A203938-1G A203938-100G A203938-500G
103817-508EA 9 USD
103817-508 103848-168 76999-428 77086-312 77086-314 77302-884 77550-302 77671-802
2-Amino-4-methylbenzoic acid 97%
2-Amino-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 329.8 °C (760 mm Hg)
Melting Pt: 174…177 °C
Flash Pt: 153.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00047853
CAS Number: 2305-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR