2-Amino-4-methylbenzamide 97%

Supplier: Ambeed
Warning

A170423-1G A170423-250MG A170423-25G
103806-490EA 32 USD
103806-490 103844-766 77081-878 77191-386 77301-052
2-Amino-4-methylbenzamide 97%
2-Amino-4-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O
Boiling Pt: 282.8 °C (760 mm Hg)
Melting Pt: 147 °C
Flash Pt: 124.823 °C
Storage Temperature: Ambient
MDL Number: MFCD00221474
CAS Number: 39549-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR