2-Amino-4-methoxypyridine 98%
Supplier: Ambeed
|
Warning
|
A106965-100G
A106965-10G
A106965-25G
A106965-500G
A106965-1G
A106965-5G
103797-100EA
5
USD
103797-100
103822-858
77107-706
77107-708
77107-710
77107-712
77309-724
77309-726
2-Amino-4-methoxypyridine 98%
2-Amino-4-methoxypyridine
Precaución: for Research Use Only
Formula:
C₆H₈N₂O Flash Pt: 110.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD07437849 CAS Number: 10201-73-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...