2-Amino-4-methoxybenzonitrile 97%

Supplier: Ambeed
Warning

A171574-10G A171574-250MG A171574-1G A171574-5G
103806-648EA 15 USD
103806-648 103844-922 76985-446 76985-448 77082-082 77082-084
2-Amino-4-methoxybenzonitrile 97%
2-Amino-4-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂O
Boiling Pt: 341.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD05149246
CAS Number: 38487-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR