2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-DL-Asp(OMe)-OH·HCl

A300086-250MG A300086-1G
103814-258EA 12 USD
103814-258 103835-408 77141-520 77305-790
2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride 97%
2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClNO₄
MW: 183.588 g/mol
Boiling Pt: 305 °C (760 mm Hg)
Melting Pt: 196…200 °C
Flash Pt: 138.2 °C
Storage Temperature: Freezer
MDL Number: MFCD00214167
CAS Number: 1835-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR