5-Iodo-o-anisidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Iodo-2-methoxyaniline, 2-Amino-4-iodoanisole, 5-Iodo-2-anisidine
A233822-10G
A233822-250MG
A233822-1G
A233822-5G
103810-912EA
30
USD
103810-912
103850-672
76771-196
76771-198
77089-648
77089-650
5-Iodo-o-anisidine 98%
5-Iodo-o-anisidine
Precaución: for Research Use Only
Formula:
C₇H₈INO MW: 249.05 g/mol Boiling Pt: 304.3 °C (760 mm Hg) Melting Pt: 85…87 °C Flash Pt: 137.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03095407 CAS Number: 77770-09-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...