2-Amino-4-fluoropyridine 97%
Supplier: Ambeed
|
Warning
|
A232517-1G
A232517-25G
A232517-5G
A232517-100G
A232517-10G
103810-838EA
7
USD
103810-838
103850-592
77089-518
77089-520
77089-522
77249-602
77249-604
2-Amino-4-fluoropyridine 97%
2-Amino-4-fluoropyridine
Precaución: for Research Use Only
Formula:
C₅H₅FN₂ Boiling Pt: 211.5 °C (760 mm Hg) Flash Pt: 81.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09260906 CAS Number: 944401-77-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...