4-Fluoroanthranilic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-fluorobenzoic acid

A236540-100G A236540-10G A236540-500G A236540-25G A236540-5G A236540-1000G
103850-880EA 6 USD
103850-880 77275-922 77275-924 77275-926 77304-056 77304-058 77748-411
4-Fluoroanthranilic acid 98%
4-Fluoroanthranilic acid

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 318.1 °C (760 mm Hg)
Melting Pt: 192…196 °C
Flash Pt: 146.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00075553
CAS Number: 446-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR