2-Amino-4-cyanopyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Aminopyrimidine-4-carbonitrile

A134947-100MG A134947-250MG A134947-1G A134947-5G
103802-076EA 36 USD
103802-076 76782-586 76782-588 77314-846 77314-848
2-Amino-4-cyanopyrimidine 98%
2-Amino-4-cyanopyrimidine

Precaución: for Research Use Only

Formula: C₅H₄N₄
MW: 120.1100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834971
CAS Number: 36314-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR