2-Aminoisonicotinonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 2-Aminopyridine-4-carbonitrile, 2-Amino-4-cyanopyridine

A222954-25G A222954-5G A222954-100G A222954-1G A222954-10G
103819-012EA 7 USD
103819-012 103849-890 77088-504 77088-506 77275-522 77303-638 77748-363
2-Aminoisonicotinonitrile 98%
2-Aminoisonicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₅N₃
MW: 119.13 g/mol
Boiling Pt: 297.7 °C (760 mm Hg)
Melting Pt: 146…148 °C
Flash Pt: 133.8 °C
Storage Temperature: Ambient
MDL Number: MFCD03791310
CAS Number: 42182-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR