2-Amino-4-chloropyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 4-chloropyrimidin-2-amine

A122503-100G A122503-10G A122503-25G A122503-500G A122503-1G A122503-5G
103800-232EA 5 USD
103800-232 76810-568 76810-570 76810-572 76810-574 77312-364 77312-366
2-Amino-4-chloropyrimidine 98%
2-Amino-4-chloropyrimidine

Precaución: for Research Use Only

Formula: C₄H₄ClN₃
MW: 129.55
Boiling Pt: 314.6 °C (760 mm Hg)
Melting Pt: 155…160 °C
Flash Pt: 144.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038021
CAS Number: 3993-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR