2-Amino-4-chloronicotinic acid 97%

Supplier: Ambeed
Warning

A216650-100MG A216650-250MG A216650-1G A216650-5G
103818-568EA 140 USD
103818-568 103849-290 76814-770 76814-772 77087-878 77087-880
2-Amino-4-chloronicotinic acid 97%
2-Amino-4-chloronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20642341
CAS Number: 605661-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR