2-Amino-4-chlorobenzamide 98%

Supplier: Ambeed
Warning

A368318-100MG A368318-10G A368318-250MG A368318-1G A368318-25G A368318-5G
103826-434EA 9 USD
103826-434 103837-448 76978-662 76978-664 76978-666 77306-796 77306-798 77306-800
2-Amino-4-chlorobenzamide 98%
2-Amino-4-chlorobenzamide

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O
MW: 170.6000 g/mol
Boiling Pt: 308.7 °C (760 mm Hg)
Flash Pt: 140.531 °C
Storage Temperature: Ambient
MDL Number: MFCD09042792
CAS Number: 5900-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR