2-Amino-4-chlorobenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A163130-100MG
A163130-250MG
A163130-1G
A163130-5G
A163130-25G
103805-690EA
23
USD
103805-690
103843-950
76985-136
76985-138
77320-036
77320-038
77890-070
2-Amino-4-chlorobenzaldehyde 97%
2-Amino-4-chlorobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₆ClNO Boiling Pt: 297.0±25.0 °C (760 mm Hg) Flash Pt: 133.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD10696881 CAS Number: 59236-37-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...