2-Amino-4-chloro-5-fluorobenzoic acid 98%

Supplier: Ambeed
Warning

A173112-100MG A173112-250MG A173112-1G A173112-5G A173112-10G
103806-790EA 43 USD
103806-790 103845-084 76974-104 76974-106 77082-310 77301-246 77671-458
2-Amino-4-chloro-5-fluorobenzoic acid 98%
2-Amino-4-chloro-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅ClFNO₂
MW: 189.57 g/mol
Boiling Pt: 350.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD05664859
CAS Number: 108288-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR