2-Amino-4-bromothiazole 95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-1,3-thiazol-2-amine

A266821-1G A266821-5G A266821-10G A266821-250MG
103812-816EA 25 USD
103812-816 103852-718 77092-424 77092-426 77166-342 77166-344
2-Amino-4-bromothiazole 95%
2-Amino-4-bromothiazole

Precaución: for Research Use Only

Formula: C₃H₃BrN₂S
MW: 179.0383 g/mol
Flash Pt: 127.7 °C
Storage Temperature: Freezer
MDL Number: MFCD06804547
CAS Number: 502145-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR