2-Amino-4-bromobenzamide 95%

Supplier: Ambeed
Warning

A138216-100MG A138216-10G A138216-1G A138216-250MG A138216-25G
103802-574EA 38 USD
103802-574 77010-232 77010-234 77298-968 77889-935 77889-936
2-Amino-4-bromobenzamide 95%
2-Amino-4-bromobenzamide

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O
Boiling Pt: 318.8 °C (760 mm Hg)
Melting Pt: 176…177 °C
Flash Pt: 146.6 °C
Storage Temperature: Ambient
MDL Number: MFCD12674809
CAS Number: 112253-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR