2-Amino-4-bromobenzaldehyde 95%

Supplier: Ambeed
Warning

A218676-1G A218676-5G A218676-100G A218676-10G A218676-250MG A218676-25G
103818-730EA 8 USD
103818-730 103849-600 77088-128 77088-130 77249-332 77249-334 77249-336 77303-502
2-Amino-4-bromobenzaldehyde 95%
2-Amino-4-bromobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₆BrNO
MW: 200.0326 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08458822
CAS Number: 59278-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR