2-Amino-4-bromo-5-fluorobenzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-5-fluoroanthranilic acid

A405892-1G A405892-5G A405892-250MG A405892-25G
103827-126EA 43 USD
103827-126 103838-166 76841-658 76841-660 77142-982 77142-984 77142-986
2-Amino-4-bromo-5-fluorobenzoic acid 95%
2-Amino-4-bromo-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
Boiling Pt: 353.6 °C (760 mm Hg)
Flash Pt: 167.6 °C
Storage Temperature: Ambient
MDL Number: MFCD08702774
CAS Number: 1374208-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR