4-Bromo-5-chloroanthranilic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-bromo-5-chlorobenzoic acid

A443500-1G A443500-25G A443500-5G A443500-10G A443500-250MG
103827-696EA 7 USD
103827-696 103838-766 76842-538 76842-540 76842-542 77252-650 77252-652
4-Bromo-5-chloroanthranilic acid 95%
4-Bromo-5-chloroanthranilic acid

Precaución: for Research Use Only

Formula: C₇H₅BrClNO₂
MW: 250.4800 g/mol
Boiling Pt: 385.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD10000554
CAS Number: 150812-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR