2-Amino-4,6-dimethylpyrimidine 97%

Supplier: Ambeed
Warning

Synonyms: 4,6-Dimethylpyrimidin-2-ylamine, 4,6-Dimethylpyrimidin-2-amine

A212791-1000G A212791-100G A212791-10G A212791-500G A212791-25G A212791-5G
103809-474EA 112 USD
103809-474 103848-930 77087-404 77112-476 77112-478 77112-480 77303-254 77303-256
2-Amino-4,6-dimethylpyrimidine 97%
2-Amino-4,6-dimethylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₉N₃
MW: 123.16 g/mol
Melting Pt: 151…153 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006102
CAS Number: 767-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR