2-Amino-4,6-dichloropyrimidine-5-carbaldehyde 95%
Supplier: Ambeed
|
Warning
|
A114791-100G
A114791-10G
A114791-25G
A114791-1G
A114791-5G
103798-474EA
9
USD
103798-474
103824-316
77284-414
77284-416
77284-418
77311-140
77311-142
2-Amino-4,6-dichloropyrimidine-5-carbaldehyde 95%
2-Amino-4,6-dichloropyrimidine-5-carbaldehyde
Precaución: for Research Use Only
Formula:
C₅H₃Cl₂N₃O Boiling Pt: 411.6 °C (760 mm Hg) Melting Pt: 208…224 °C Flash Pt: 202.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03001242 CAS Number: 5604-46-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...