2-Amino-4,6-dichloropyrimidine 98%
Supplier: Ambeed
|
Warning
|
A134107-1000G
A134107-100G
A134107-10G
A134107-500G
A134107-5G
A134107-25G
77272-120EA
123.6
USD
77272-120
77272-122
77272-124
77272-126
77298-688
77314-694
2-Amino-4,6-dichloropyrimidine 98%
2-Amino-4,6-dichloropyrimidine
Precaución: for Research Use Only
Formula:
C₄H₃Cl₂N₃ MW: 163.99 g/mol Boiling Pt: 348.5 °C (760 mm Hg) Melting Pt: 219…222 °C Flash Pt: 164.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006090 CAS Number: 56-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...