2-Amino-4,5-dibromobenzoic acid 97%

Supplier: Ambeed
Warning

A160479-100MG A160479-250MG A160479-1G A160479-5G
103805-384EA 39 USD
103805-384 103843-616 77247-650 77247-652 77319-554 77319-556
2-Amino-4,5-dibromobenzoic acid 97%
2-Amino-4,5-dibromobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅Br₂NO₂
MW: 294.92 g/mol
Boiling Pt: 383.9 °C (760 mm Hg)
Melting Pt: 228…229 °C
Storage Temperature: Ambient
MDL Number: MFCD00017100
CAS Number: 75057-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR