2-Amino-4,5-bis(2-methoxyethoxy)benzonitrile 97%

Supplier: Ambeed
Warning

A148074-10G A148074-250MG A148074-1G A148074-5G
103803-874EA 11 USD
103803-874 103841-988 76823-728 76823-730 77299-634 77299-636
2-Amino-4,5-bis(2-methoxyethoxy)benzonitrile 97%
2-Amino-4,5-bis(2-methoxyethoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₄
MW: 266.29 g/mol
Boiling Pt: 445.5±45.0 °C (760 mm Hg)
Melting Pt: 73…77 °C
Flash Pt: 223.2 °C
Storage Temperature: Ambient
MDL Number: MFCD16619377
CAS Number: 950596-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR