2-Amino-4-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Trifluoromethyl)pyridin-2-amine, 4-(Trifluoromethyl)-2-pyridylamine

A297024-1G A297024-25G A297024-5G A297024-100G A297024-10G A297024-500G
103814-120EA 6 USD
103814-120 103835-262 76837-946 76837-948 76837-950 77099-444 77099-446 77099-448
2-Amino-4-(trifluoromethyl)pyridine 98%
2-Amino-4-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₅F₃N₂
Melting Pt: 70…74 °C
Flash Pt: 87.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00077455
CAS Number: 106447-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR