2-Amino-4-(trifluoromethyl)benzonitrile 98%

Supplier: Ambeed
Warning

A119322-10G A119322-25G A119322-1G A119322-5G
103799-752EA 8 USD
103799-752 76766-378 76766-380 77311-874 77311-876
2-Amino-4-(trifluoromethyl)benzonitrile 98%
2-Amino-4-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₅F₃N₂
Melting Pt: 88…89 °C
Flash Pt: 120.8 °C
Storage Temperature: Ambient
MDL Number: MFCD04974124
CAS Number: 1483-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR