4-(Trifluoromethyl)anthranilic acid 98%

Supplier: Ambeed
Warning

A109350-100G A109350-10G A109350-25G A109350-5G
103823-294EA 23 USD
103823-294 77233-452 77233-454 77233-456 77310-110
4-(Trifluoromethyl)anthranilic acid 98%
4-(Trifluoromethyl)anthranilic acid

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
Boiling Pt: 296 °C (760 mm Hg)
Melting Pt: 175…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00519334
CAS Number: 402-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR