2-Amino-3-nitropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Nitropyridin-2-ylamine, 3-Nitropyridin-2-amine, 3-Nitro-2-pyridylamine

A171849-5G A171849-100G A171849-10G A171849-500G A171849-25G
103844-950EA 9 USD
103844-950 77082-122 77286-898 77286-900 77286-902 77301-168
2-Amino-3-nitropyridine 98%
2-Amino-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
Boiling Pt: 299.1 °C (760 mm Hg)
Melting Pt: 163…165 °C
Flash Pt: 167 °C
Storage Temperature: Ambient
MDL Number: MFCD00006314
CAS Number: 4214-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR