2-Amino-3-nitrophenol 98%

Supplier: Ambeed
Warning

A110004-100G A110004-10G A110004-1G A110004-25G A110004-500G A110004-5G A110004-1000G
103823-410EA 8 USD
103823-410 77283-920 77283-922 77283-924 77283-926 77283-928 77283-930 77747-811
2-Amino-3-nitrophenol 98%
2-Amino-3-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
Boiling Pt: 315.8 °C (760 mm Hg)
Melting Pt: 212…213 °C
Flash Pt: 144.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00010875
CAS Number: 603-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR