2-Amino-6-methyl-3-nitropyridine 95%

Supplier: Ambeed
Warning

Synonyms: 6-Methyl-3-nitro-2-pyridylamine, 6-Methyl-3-nitropyridin-2-amine, 2-Amino-3-nitro-6-picoline, 2-Amino-3-nitro-6-picoline

A167440-1G A167440-5G A167440-100G A167440-10G
103806-186EA 9 USD
103806-186 103844-448 77081-466 77081-468 77110-894 77110-896
2-Amino-6-methyl-3-nitropyridine 95%
2-Amino-6-methyl-3-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
Boiling Pt: 309.1 °C (760 mm Hg)
Melting Pt: 147…157 °C
Flash Pt: 140.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00047443
CAS Number: 21901-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR