2-Amino-3-nitro-5-fluoropyridine 98%

Supplier: Ambeed
Warning

A225088-250MG A225088-1G
103819-156EA 16 USD
103819-156 103850-048 77000-036 77088-720
2-Amino-3-nitro-5-fluoropyridine 98%
2-Amino-3-nitro-5-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₄FN₃O₂
Flash Pt: 129.3 °C
Storage Temperature: Ambient
MDL Number: MFCD04972413
CAS Number: 212268-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR