2-Amino-m-cresol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-6-methylaniline, 2-Amino-3-hydroxytoluene, 2-Amino-3-methylphenol, 1-Amino-2-hydroxy-6-methylbenzene

A105706-10G A105706-250MG A105706-25G A105706-1G A105706-5G
103796-928EA 10 USD
103796-928 103822-666 76994-892 76994-894 76994-896 77283-390 77309-530
2-Amino-m-cresol 97%
2-Amino-m-cresol

Precaución: for Research Use Only

Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 240 °C (760 mm Hg)
Melting Pt: 149…152 °C
Flash Pt: 98.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00075082
CAS Number: 2835-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR