3-Methylanthranil acid 98%

Supplier: Ambeed
Warning

A113799-100G A113799-10G A113799-25G A113799-500G A113799-5G A113799-1000G
103808-108EA 147 USD
103808-108 77271-082 77271-084 77271-086 77271-088 77271-090 77297-268
3-Methylanthranil acid 98%
3-Methylanthranil acid

Precaución: for Research Use Only

Formula: C₈H₉NO₂
Boiling Pt: 315.1 °C (760 mm Hg)
Melting Pt: 174…177 °C
Flash Pt: 144.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00007745
CAS Number: 4389-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR