2-Amino-3-methoxypyridine 98%

Supplier: Ambeed
Warning

A433092-1G A433092-25G A433092-5G A433092-100G A433092-10G
103827-542EA 10 USD
103827-542 103838-608 76842-312 76842-314 76842-316 76990-512 76990-514
2-Amino-3-methoxypyridine 98%
2-Amino-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MW: 124.1400 g/mol
Boiling Pt: 235 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07374874
CAS Number: 10201-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR