2-Amino-3-methoxyphenol 97%

Supplier: Ambeed
Warning

A311382-100MG A311382-250MG A311382-5G A311382-1G
103814-664EA 134 USD
103814-664 103835-852 76828-000 76828-002 76838-646 77306-054
2-Amino-3-methoxyphenol 97%
2-Amino-3-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MW: 139.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12963875
CAS Number: 40925-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR