2-Amino-3-methoxybenzonitrile 96%

Supplier: Ambeed
Warning

A115487-10G A115487-250MG A115487-25G A115487-1G A115487-5G
103798-572EA 45 USD
103798-572 103824-428 76995-500 76995-502 76995-504 77311-244 77311-246
2-Amino-3-methoxybenzonitrile 96%
2-Amino-3-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂O
Boiling Pt: 286.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09744022
CAS Number: 148932-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR