3-Methoxyanthranilic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-m-anisic acid, 3-Methoxyanthranilic acid, o-Anisidine-6-carboxylic acid, 2-Amino-3-methoxybenzoic acid

A282625-1G A282625-25G A282625-5G A282625-10G A282625-100G
103813-596EA 6 USD
103813-596 103853-548 76837-246 76837-248 76837-250 77250-528 77746-513
3-Methoxyanthranilic acid 97%
3-Methoxyanthranilic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Boiling Pt: 333.9 °C (760 mm Hg)
Melting Pt: 169…170 °C
Flash Pt: 155.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00075178
CAS Number: 3177-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR