2-Amino-3-methoxy-4-fluorobenzoic acid 98%

Supplier: Ambeed
Warning

A156931-100MG A156931-250MG A156931-1G A156931-5G
103804-938EA 55 USD
103804-938 103843-140 76973-342 76973-344 77300-224 77318-946
2-Amino-3-methoxy-4-fluorobenzoic acid 98%
2-Amino-3-methoxy-4-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₈FNO₃
MW: 185.15 g/mol
Boiling Pt: 346.9 °C (760 mm Hg)
Flash Pt: 163.572 °C
Storage Temperature: Ambient
MDL Number: MFCD14706679
CAS Number: 126480-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR