DL-Cysteine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-mercaptopropanoic acid, H-DL-Cys-OH, (±)-Cysteine

A181229-25G A181229-5G A181229-10G A181229-100G A181229-500G
103845-958EA 8 USD
103845-958 77083-480 77083-482 77096-860 77685-419 77685-420
DL-Cysteine 95%
DL-Cysteine

Precaución: for Research Use Only

Formula: C₃H₇NO₂S
MW: 121.16
Boiling Pt: 254
Melting Pt: 115
Density: 1.441
Flash Pt: 131.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00004881
CAS Number: 3374-22-9
Merck Index: 12,02850

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR