2-Amino-3-pyridinemethanol 97%

Supplier: Ambeed
Warning

A118031-10G A118031-250MG A118031-25G A118031-1G A118031-5G
103798-978EA 8 USD
103798-978 103824-872 76995-588 76995-590 76995-592 77311-658 77311-660
2-Amino-3-pyridinemethanol 97%
2-Amino-3-pyridinemethanol

Precaución: for Research Use Only

Formula: C₆H₈N₂O
Flash Pt: 147.7 °C
Storage Temperature: Ambient
MDL Number: MFCD05663510
CAS Number: 23612-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR