2-Amino-3-hydroxybenzoic acid 98%

Supplier: Ambeed
Warning

A105554-100G A105554-10G A105554-1G A105554-25G
103796-900EA 19 USD
103796-900 103822-634 77188-764 77188-766 77283-364 77745-799
2-Amino-3-hydroxybenzoic acid 98%
2-Amino-3-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₃
Boiling Pt: 356.8 °C (760 mm Hg)
Melting Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00007700
CAS Number: 548-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR